missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organochlorine Pesticides Mix, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Aldrin | 309-00-2 | 2000μg/mL |
| alpha-BHC | 319-84-6 | 2000μg/mL |
| beta-BHC | 319-85-7 | 2000μg/mL |
| delta-BHC | 319-86-8 | 2000μg/mL |
| Dieldrin | 60-57-1 | 2000μg/mL |
| Endosulfan I | 959-98-8 | 2000μg/mL |
| Endosulfan II | 33213-65-9 | 2000μg/mL |
| Endosulfan sulfate | 1031-07-8 | 2000μg/mL |
| Endrin | 72-20-8 | 2000μg/mL |
| Endrin aldehyde | 7421-93-4 | 2000μg/mL |
| Endrin ketone | 53494-70-5 | 2000μg/mL |
| gamma-BHC | 58-89-9 | 2000μg/mL |
| Heptachlor | 76-44-8 | 2000μg/mL |
| Heptachlor epoxide (Isomer B) | 1024-57-3 | 2000μg/mL |
| Methoxychlor | 72-43-5 | 2000μg/mL |
| p,p'-DDD | 72-54-8 | 2000μg/mL |
| p,p'-DDE | 72-55-9 | 2000μg/mL |
| p,p'-DDT | 50-29-3 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Organochlorine Pesticides Mix |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| Methods | For US EPA Methods 625, 8270C |
| Analyte or Component Names | Aldrin; alpha-BHC; beta-BHC; p,p'-DDD; p,p'-DDE; delta-BHC; Dieldrin; p |
| Technique | US EPA Methods 625 and 8270 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Element | Aldrin, alpha-BHC, beta-BHC, p,p'-DDD, p,p'-DDE, delta-BHC, Dieldrin |
| Solvent or Matrix | Benzene |
| Show More |
Safety and Handling
Product Identifier
- Organochlorine Pesticides Mix
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 3
- Aspiration toxicity Category 1
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity on or via lactation
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H362-May cause harm to breast-fed children.
- H372-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : benzene, delta-BHC,dieldrin, heptachlor, heptachlor epoxide, 1,1,1-trichloro-2,2-bis(4-chlorophenyl
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction