Learn More
Analyte Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 608-A
Additional Details : Weight : 0.00100kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Endosulfan sulfate | 1031-07-8 | 600μg/mL |
Endrin aldehyde | 7421-93-4 | 600μg/mL |
p,p'-DDD | 72-54-8 | 600μg/mL |
p,p'-DDT | 50-29-3 | 600μg/mL |
Dieldrin | 60-57-1 | 200μg/mL |
Endosulfan I | 959-98-8 | 200μg/mL |
Endosulfan II | 33213-65-9 | 200μg/mL |
Endrin | 72-20-8 | 200μg/mL |
p,p'-DDE | 72-55-9 | 200μg/mL |
Aldrin | 309-00-2 | 100μg/mL |
alpha-BHC | 319-84-6 | 100μg/mL |
beta-BHC | 319-85-7 | 100μg/mL |
delta-BHC | 319-86-8 | 100μg/mL |
gamma-BHC | 58-89-9 | 100μg/mL |
Heptachlor | 76-44-8 | 100μg/mL |
Heptachlor epoxide (Isomer B) | 1024-57-3 | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Analyte Mix | |
For US EPA Method 608 | |
Methanol | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
2mL amber prescored glass ampule | |
100μg/mL Aldrin; 100μg/mL α-BHC; 100μg/mL β-BHC; 100μg/mL γ-BHC; 100μg/mL δ-BHC; 600μg/mL; 4,4'-DDD; 200μg/mL 4,4'-DDE; 600μg/mL 4 | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Safety and Handling
- 608 STD MIX
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol
Product intended for laboratory use only