missing translation for 'onlineSavingsMsg'
Learn More

Organochlorine Pesticides Mix B, SPEX CertiPrep™

Product Code. 15285786 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15285786 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15285786 Supplier SPEX Certiprep Supplier No. 5252PB
  • ONLINE EXCLUSIVE PRICE
    £167.00
    £148.44 / 1mL
    Save £18.56
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Alachlor15972-60-8200μg/mL
alpha-Chlordane5103-71-9200μg/mL
Bladex21725-46-2200μg/mL
Chlorobenzilate510-15-6200μg/mL
Chloroneb2675-77-6200μg/mL
Chlorothalonil1897-45-6200μg/mL
Chlorpyrifos2921-88-2200μg/mL
gamma-Chlordane5103-74-2200μg/mL
Hexachlorobenzene118-74-1200μg/mL
Methyl dacthal1861-32-1200μg/mL
Permethrin (c=39%;t=59%)52645-53-1200μg/mL
Propachlor1918-16-7200μg/mL
Terrazole2593-15-9200μg/mL
trans-Nonachlor39765-80-5200μg/mL
Trifluralin1582-09-8200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Organochlorine Pesticides Mix B
Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA Method 525.2
Analyte or Component Names Alachlor; Bladex; alpha-Chlordane; Chloroneb; Chlorobenzilate; Chlorothalonil; Chlorpyrifos; gamma-Chlordane; Hexachlorobenzene; Methyl dacthal; trans-Nonachlor; Permethrin (t=55%;c=44%); Propachlor; Terrazole; Trifluralin
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Element Alachlor, Bladex, alpha-Chlordane, Chloroneb, Chlorobenzilate, Chlorothalonil, Chlorpyrifos, gamma-Chlordane, Hexachlorobenzene, Methyl Dacthal, trans-Nonachlor, Permethrin, Propachlor, Terrazole, Trifluralin
Solvent or Matrix Acetone
Quantity 1 mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Organochlorine pest MixB
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetone, chlorothalonil, chlorpyrifos, trifluralin

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.