missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MethoCult™ SF H4436
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- MethoCult™ SF H4436 is a complete serum-free methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples
- MethoCult™ SF H4436 is formulated to support optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage, megakaryocyte progenitors (CFU-GEMM) under defined serum-free conditions
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction