missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MethoCult™ M3630
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- MethoCult™ M3630 is optimized for the growth and enumeration of mouse pre-B lymphoid progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow and Whitlock-Witte long-term cultures
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction