missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MethoCult™ M3334

Product Code. 17161026
Encompass
Change view
Click to view available options
Quantity:
90 mL
Unit Size:
90mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17161026 90 mL 90mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161026 Supplier STEMCELL Technologies Supplier No. 03334

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methylcellulose-based medium with EPO (without other cytokines) for mouse cells

  • MethoCult™ M3334 is optimized for the growth and enumeration of erythroid progenitor cells (late BFU-E and CFU-E) in colony-forming unit (CFU) assays of mouse bone marrow, spleen, and fetal liver cells
  • This formulation contains erythropoietin (EPO) but does not contain other recombinant cytokines

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 90 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.