missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MethoCult™ H4534 Classic Without EPO, Pre-aliquoted

Product Code. 15549525
Encompass
Change view
Click to view available options
Quantity:
24 x 3 mL
Unit Size:
72mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15549525 24 x 3 mL 72mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15549525 Supplier STEMCELL Technologies Supplier No. 04544

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-aliquoted methylcellulose-based medium with recombinant cytokines (without EPO) for human cells

  • MethoCult™ H4534 Classic Without EPO (MethoCult™ GF H4534) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples
  • MethoCult™ H4534 Classic Without EPO is formulated to support the optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M)

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 24 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.