missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MethoCult™ H4100

Product Code. 17188211
Encompass
Click to view available options
Quantity:
40 mL
Unit Size:
40mL
This item is not returnable. View return policy

Product Code. 17188211

Brand: STEMCELL Technologies 04100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Base methylcellulose medium for human cells

  • MethoCult™ H4100 is an incomplete medium that contains 2.6% methylcellulose in Iscove's MDM
  • MethoCult™ H4100 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples, when the appropriate growth factors and supplements are added
  • This formulation does not contain serum or cytokines and can also be used as a base for cloning cell lines in semi-solid media

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 40 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.