missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MethoCult™ GF M3434, Pre-aliquoted

Product Code. 17158081
Encompass
Change view
Click to view available options
Quantity:
24 x 3 mL
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17158081 24 x 3 mL Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17158081 Supplier STEMCELL Technologies Supplier No. 03444

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-aliquoted methylcellulose-based medium with recombinant cytokines (including EPO) for mouse cells

  • MethoCult™ GF M3434 is optimized for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver cells
  • MethoCult™ GF M3434 has been formulated to support optimal growth of primitive erythroid progenitor cells (BFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multi-potential granulocyte, erythroid, macrophage, megakaryocyte progenitor cells (CFU-GEMM)
  • This formulation is compatible with STEMvision™ software for automated colony counting of mouse bone marrow CFU assays

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 24 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.