missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Methocarbamol-d3 (methoxy-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Methocarbamol-d3 |
| CAS | 1346600-86-9 |
| Molecular Formula | C11H12D3NO5 |
| Quantity | 0.005 g |
| Synonym | 3-(2-Methoxyphenoxy)-1,2-propanediol-1-carbamate-d3, 3-(o-Methoxyphenoxy) -2-hydroxypropyl Carbamate-d3, AHR 85-d3, Avetil-d3, Delaxin-d3, Etroflex-d3, Lumirelax-d3, Miolaxene-d3, Neuraxin-d3, Relestrid-d3, Robinax-d3; |
| InChI Formula | InChI=1 S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13 H,6-7H2,1H3,(H2,12,14)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccccc1OCC(O)COC(=O)N |
| IUPAC Name | [2-hydroxy-3-[2-(trideuteriomethoxy)phenoxy]propyl] carbamate |
| Molecular Weight (g/mol) | 244.26 |
| Formula Weight | 244.1139 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction