missing translation for 'onlineSavingsMsg'
Learn More

Methionyl tRNA synthetase Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18413327 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413327 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413327 Supplier Novus Biologicals Supplier No. NBP222313AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Methionyl tRNA synthetase Polyclonal antibody specifically detects Methionyl tRNA synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methionyl tRNA synthetase
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias cytosolic methionyl-tRNA synthetase, EC 6.1.1.10, FLJ35667, methionine tRNA ligase 1, cytoplasmic, Methionine--tRNA ligase, methionyl-tRNA synthetase, methionyl-tRNA synthetase, cytoplasmic, METRS, MRS, MTRNS
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 650 to 700 of human Methionyl-tRNA Synthetase using the numbering given in entry NP_004981.2 (GeneID 4141).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4141
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.