missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionyl tRNA synthetase Antibody, Alexa Fluor™ 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Methionyl tRNA synthetase Polyclonal antibody specifically detects Methionyl tRNA synthetase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Methionyl tRNA synthetase |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | cytosolic methionyl-tRNA synthetase, EC 6.1.1.10, FLJ35667, methionine tRNA ligase 1, cytoplasmic, Methionine--tRNA ligase, methionyl-tRNA synthetase, methionyl-tRNA synthetase, cytoplasmic, METRS, MRS, MTRNS |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 175 to 225 of human Methionyl-tRNA Synthetase using the numbering given in entry NP_004981.2 (GeneID 4141). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?