missing translation for 'onlineSavingsMsg'
Learn More

Methionine Aminopeptidase 2/METAP2 Rabbit anti-Human, Mouse, Rat, Clone: 5U2F4, Novus Biologicals™

Product Code. 18301525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18301525 20 μg 20µL
18346724 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18301525 Supplier Novus Biologicals Supplier No. NBP31619320UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Methionine Aminopeptidase 2/METAP2 Monoclonal antibody specifically detects Methionine Aminopeptidase 2/METAP2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methionine Aminopeptidase 2/METAP2
Applications Western Blot
Classification Monoclonal
Clone 5U2F4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 380-478 of human Methionine Aminopeptidase 2/METAP2 (P50579).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10988
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.