missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Aminopeptidase 2/METAP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£248.00 - £565.00
Specifications
| Antigen | Methionine Aminopeptidase 2/METAP2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656676
|
Novus Biologicals
NBP3-05513-100ul |
100 μg |
£565.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646977
|
Novus Biologicals
NBP3-05513-25ul |
25 μg |
£248.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methionine Aminopeptidase 2/METAP2 Polyclonal antibody specifically detects Methionine Aminopeptidase 2/METAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Methionine Aminopeptidase 2/METAP2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| PBS, pH 7.2, containing 40% glycerol | |
| 10988 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SGSHLNGDLDPDDREEGAASTAEEAAKKKRRKKKKSKGPSAAGEQEPDKESGASVDEVARQLERSALEDKERDEDDEDGDGDGDGATGKKKK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title