missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Aminopeptidase 1D/MAP1D Antibody, mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Methionine Aminopeptidase 1D/MAP1D Polyclonal antibody specifically detects Methionine Aminopeptidase 1D/MAP1D in Human samples. It is validated for ELISA
Specifications
Specifications
| Purification Method | Antigen and protein A Affinity-purified |
| Research Discipline | Immunology |
| Gene ID (Entrez) | 254042 |
| Target Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?