missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Aminopeptidase 1D/MAP1D Antibody (008), DyLight 650, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Methionine Aminopeptidase 1D/MAP1D Monoclonal antibody specifically detects Methionine Aminopeptidase 1D/MAP1D in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Methionine Aminopeptidase 1D/MAP1D |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 008 |
| Conjugate | DyLight 650 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.4.11.18, MAP1DCDS of metAP-3 within PCR fragment, Metap1l, methionine aminopeptidase 1D, mitochondrial, methionyl aminopeptidase type 1D (mitochondrial), Methionyl aminopeptidase type 1D, mitochondrial |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Methionine Aminopeptidase 1D/MAP1D (NP_954697.1; Arg 44-Ala 335). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?