missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Aminopeptidase 1/METAP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18418JF549
This item is not returnable.
View return policy
Description
Methionine Aminopeptidase 1/METAP1 Polyclonal antibody specifically detects Methionine Aminopeptidase 1/METAP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Methionine Aminopeptidase 1/METAP1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp781C0419, EC 3.4.11.18, KIAA0094MAP 1, MAP1A, MetAP 1, MetAP1A, methionine aminopeptidase 1, methionyl aminopeptidase 1, Peptidase M 1 | |
| Recognizes region between residues Between Between 336 and 386.. | |
| 0.1 mL | |
| metabolism, Proteases & Other Enzymes | |
| 23173 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction