missing translation for 'onlineSavingsMsg'
Learn More

Methimazole, TRC

Product Code. 30481438
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
Product Code. Quantity unitSize
30481438 50 mg 50mg
30469693 1 g 1g
30464462 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 30481438

Brand: Toronto Research Chemicals TRCM26030050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-56-0
Formula Weight 114.0252
InChI Formula InChI=1S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3H,1H3,(H,5,7)
IUPAC Name 3-methyl-1H-imidazole-2-thione
Molecular Formula C4 H6 N2 S
Molecular Weight (g/mol) 114.17
Percent Purity >95%
SMILES CN1C=CNC1=S
Synonym 4-Imidazoline-2-thione, 1-methyl- (6CI,7CI), Imidazole-2-thiol, 1-methyl- (8CI), 1,3-Dihydro-1-methyl-2H-imidazole-2-thione, 1-Methyl-1,3-dihydroimidazole-2-thione, 1-Methyl-1H-imidazole-2-thiol, 1-Methyl-2-mercapto-1H-imidazole, 1-Methyl-2-mercaptoimidazole, 1-Methyl-4-imidazoline-2-thione, 1-Methylimidazole-2(3H)-thione, 1-Methylimidazole-2-thiol, 1-Methylimidazole-2-thione, 2-Mercapto-1-methyl-1H-imidazole, 2-Mercapto-1-methylimidazole, 2-Mercapto-N-methylimidazole, Basolan, Danantizol, Favistan, Frentirox, MMI, Mercaptazole, Mercazole, Mercazolyl, Metazolo, Methimazol, Methimazole, Methylmercaptoimidazole, Metizol, Metothyrin, Metothyrine, Metotirin, N-Methyl-2-mercaptoimidazole, N-Methylimidazolethiol, NSC 38608, Strumazol, Tapazole, Thacapzol, Thiamazol, Thiamazole, Thycapzol, Thymidazol, Thymidazole, Thyrozol
Chemical Name or Material Methimazol
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.