missing translation for 'onlineSavingsMsg'
Learn More

Methimazole, TRC

Product Code. 30469693
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469693 1 g 1g
30481438 50 mg 50mg
30464462 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469693 Supplier Toronto Research Chemicals Supplier No. TRCM2603001G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-56-0
Molecular Formula C4 H6 N2 S
Molecular Weight (g/mol) 114.17
Synonym 4-Imidazoline-2-thione, 1-methyl- (6CI,7CI), Imidazole-2-thiol, 1-methyl- (8CI), 1,3-Dihydro-1-methyl-2H-imidazole-2-thione, 1-Methyl-1,3-dihydroimidazole-2-thione, 1-Methyl-1H-imidazole-2-thiol, 1-Methyl-2-mercapto-1H-imidazole, 1-Methyl-2-mercaptoimidazole, 1-Methyl-4-imidazoline-2-thione, 1-Methylimidazole-2(3H)-thione, 1-Methylimidazole-2-thiol, 1-Methylimidazole-2-thione, 2-Mercapto-1-methyl-1H-imidazole, 2-Mercapto-1-methylimidazole, 2-Mercapto-N-methylimidazole, Basolan, Danantizol, Favistan, Frentirox, MMI, Mercaptazole, Mercazole, Mercazolyl, Metazolo, Methimazol, Methimazole, Methylmercaptoimidazole, Metizol, Metothyrin, Metothyrine, Metotirin, N-Methyl-2-mercaptoimidazole, N-Methylimidazolethiol, NSC 38608, Strumazol, Tapazole, Thacapzol, Thiamazol, Thiamazole, Thycapzol, Thymidazol, Thymidazole, Thyrozol
IUPAC Name 3-methyl-1H-imidazole-2-thione
SMILES CN1C=CNC1=S

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3H,1H3,(H,5,7)
Formula Weight 114.0252
Percent Purity >95%
Chemical Name or Material Methimazol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.