missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methicillin Resistant Staphylococcus Aureus Antibody (332/423), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54428JF549
This item is not returnable.
View return policy
Description
Methicillin Resistant Staphylococcus Aureus Monoclonal antibody specifically detects Methicillin Resistant Staphylococcus Aureus in S. aureus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Methicillin Resistant Staphylococcus Aureus | |
| Monoclonal | |
| Janelia Fluor 549 | |
| MRSA, s aureus, S. aureus, staphylococcus aureus methicillin-resistant | |
| Methicillin-resistant Staphylococcus aureus (MRSA) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| ELISA, Immunofluorescence | |
| 332/423 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| S. aureus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction