missing translation for 'onlineSavingsMsg'
Learn More

Methdilazine Hydrochloride, TRC

Product Code. 30481535
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481535 500 mg 500mg
30468379 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481535 Supplier Toronto Research Chemicals Supplier No. TRCM260100500MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1229-35-2
Molecular Formula C18 H20 N2 S . Cl H
Molecular Weight (g/mol) 332.89
Synonym 10H-Phenothiazine, 10-[(1-methyl-3-pyrrolidinyl)methyl]-, hydrochloride (1:1), 10H-Phenothiazine, 10-[(1-methyl-3-pyrrolidinyl)methyl]-, monohydrochloride (9CI), Phenothiazine, 10-[(1-methyl-3-pyrrolidinyl)methyl]-, monohydrochloride (8CI), Dilosyn, Disyncran, Methdilazine hydrochloride, Methdilazine monohydrochloride, NSC 169091, Tacaryl, Tacaryl hydrochloride
IUPAC Name 10-[(1-methylpyrrolidin-3-yl)methyl]phenothiazine;hydrochloride
SMILES Cl.CN1CCC(CN2c3ccccc3Sc4ccccc24)C1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H20N2S.ClH/c1-19-11-10-14(12-19)13-20-15-6-2-4-8-17(15)21-18-9-5-3-7-16(18)20;/h2-9,14H,10-13H2,1H3;1H
Formula Weight 332.1114
Chemical Name or Material Methdilazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.