missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methazolamide, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
Description
Methazolamide (L584601) is a sulfonamide derivative used as a carbonic anhydrase inhibitor with a Ki of 14 nM for human carbonic anhydrase II. Methazolamide, an intraocular pressure-lowering agent, reduces intraocular pressure elevations associated with glaucoma and other ocular disorders.
Specifications
Specifications
| Chemical Name or Material | Methazolamide |
| CAS | 554-57-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C5H8N4O3S2 |
| Quantity | 1 g |
| Synonym | L584601 |
| Solubility Information | DMSO : ≥ 50 mg/mL (211.62 mM) |
| SMILES | CC(/N=C1SC(S(=O)(N)=O)=NN/1C)=O |
| Molecular Weight (g/mol) | 236.27 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction