missing translation for 'onlineSavingsMsg'
Learn More

Methazolamide, MedChemExpress

Product Code. 16478366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478366 100 mg 100mg
16462229 1 g 1g
16420998 5 g 5g
16412176 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478366 Supplier Medchem Express Supplier No. HYB0553
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methazolamide (L584601) is a sulfonamide derivative used as a carbonic anhydrase inhibitor with a Ki of 14 nM for human carbonic anhydrase II. Methazolamide, an intraocular pressure-lowering agent, reduces intraocular pressure elevations associated with glaucoma and other ocular disorders.

Methazolamide (L584601) is a sulfonamide derivative used as a carbonic anhydrase inhibitor with a Ki of 14 nM for human carbonic anhydrase II. Methazolamide, an intraocular pressure-lowering agent, reduces intraocular pressure elevations associated with glaucoma and other ocular disorders.

Specifications

Chemical Name or Material Methazolamide
CAS 554-57-4
Color White
Purity Grade Notes Research
Molecular Formula C5H8N4O3S2
Quantity 100 mg
Synonym L584601
Solubility Information DMSO : ≥ 50 mg/mL (211.62 mM)
SMILES CC(/N=C1SC(S(=O)(N)=O)=NN/1C)=O
Molecular Weight (g/mol) 236.27
Formula Weight 236.27
Percent Purity 99.43%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.