missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Semiconductor Grade, 99.9% min

Product Code. 11383557
Change view
Click to view available options
Quantity:
100 mL
1 L
4 x 1 L
Unit Size:
1 L
100mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383557 100 mL 100mL
11393557 1 L 1 L
11373557 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11383557 Supplier Thermo Scientific Alfa Aesar Supplier No. 019393.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Acidity 0.3meq/g, Alkalinity 0.1meq/g, Evaporation residue 5ppm, H2O (by Karl Fischer titrn) 0.05%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.5ppm | CAS: 67-56-1 | CH4O | 32.04 g/mol

Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Appearance Colorless liquid
Density 0.791g/cm3
Quantity 100 mL
Ignition Point 455°C (851°F)
Physical Form Liquid
Assay Percent Range 99.9% min.
Vapor Pressure 128 hPa (96mm Hg) at 20°C
Formula Weight 32.04
Grade Semiconductor
Chemical Name or Material Methanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.