missing translation for 'onlineSavingsMsg'
Learn More

Methanol (Purge and Trap Suitable for Volatile Organic Residue Analysis), Fisher Chemical™

Product Code. 30117586 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
30117586 500 mL Amber Glass 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30117586 Supplier Fisher Chemical Supplier No. A453500
  • ONLINE EXCLUSIVE PRICE
    £52.65
    £51.70 / 500mL
    Save £0.95
    1% Off
 Restricted Item
Estimated Shipment: 21-05-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Packaged under nitrogen. Submicron filtered

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791g/cm³
Quantity 500 mL
Flash Point 12°C (53.6°F)
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation 1ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0002mEq/g max.
Linear Formula CH3OH
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
Water 0.3% max. (Karl Fischer)
Formula Weight 32.04
Percent Purity ≥99.9%
Grade Residue Analysis
Chemical Name or Material Methanol, Purge and trap suitable for volatile organic residue analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.