missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Purge and Trap, ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. 15554055 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15554055 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15554055 Supplier JT Baker Supplier No. 907702

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High purity solvents and salts developed for organic residue extraction/concentration procedures

A group of ultra high purity solvents and salts developed for organic residue extraction/concentration procedures.

Specifications

Melting Point -98°C
CAS 67-56-1
Color Colorless
Quantity 1 L
Explosion Limit 5.5 to 44 vol%
Flash Point 11°C
Ignition Point 455°C
Odor Alcohol-like
Physical Form Liquid
Titratable Acid ≤0.3μeq/g
Molecular Formula CH4O
MDL Number MFCD00004595
UN Number 1230
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Boiling Point 64.7°C
Solubility Information Fully miscible in water
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Percent Purity ≥99.9%
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Methanol, Purge and Trap
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.