missing translation for 'onlineSavingsMsg'
Learn More

Methanol, LC/MS grade, Thermo Scientific

Brand:  Thermo Scientific T004561000

42.39 GBP valid until 2026-10-31
BEST PRICE on Promo! Use promo code "27992" to get your promotional price.



 View more versions of this product

Product Code. 30280931

  • ONLINE EXCLUSIVE PRICE
    £175.00
    £47.10 / 1 L
    Save £127.90
    73% Off
View all promotions
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contain: Methanol

SDS
Documents

Documents

Certificates
Promotions

Promotions

Promo
Expires: 2026-10-31
Big Savings for Better Separation
Promo Code: 27992
Save up to 60% on chromatography consumables.

Applying Promo and Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.