missing translation for 'onlineSavingsMsg'
Learn More

Methanol, HPLC Plus, ≥99.9%; Honeywell™ Riedel-de Haën™

Product Code. 15676100
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15676100 1 L Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15676100 Supplier Honeywell-Chromasolv Supplier No. 6463771L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™ Plus, for HPLC

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791g/mL at 25°C, 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.36
Ignition Point 725°C
Packaging Poly Bottle
Linear Formula CH3OH
Vapor Pressure 410mmHg (50°C), 97.68mmHg (20°C)
UN Number UN1230
Beilstein 1098229
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade HPLC Plus
Chemical Name or Material Methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.