missing translation for 'onlineSavingsMsg'
Learn More

Methanol, for HPLC, Gradient Grade, ACS, Honeywell Burdick & Jackson™

Product Code. 16625893
Click to view available options
Quantity:
7 L
30 L
45 L
395 L
Unit Size:
30 L
395 L
45 L
7 L
This item is not returnable. View return policy

Product Code. 16625893

Brand: Honeywell Burdick & Jackson 6572745L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
CAS Max % 1
Density 0.79 g/cm3 (20°C)
Boiling Point 64°C to 65°C (1013 hPa)
Quantity 45 L
Flash Point 11°C
Chemical Name or Material Methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.