Learn More
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
CH4O, CAS Number-67-56-1, methyl hydroxide, methylol, pyroxylic spirit, carbinol, wood spirit, wood naphtha, wood alcohol, colonial spirit, columbian spirit, methyl alcohol, 1L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042
Brand: Fisher Chemical M/4050/08
Code : 25
Additional Details : CAS Number : 67-56-1 Weight : 0.39600kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Packaging | Amber glass bottle |
---|---|
Quantity | 500mL |
Description
CH4O, CAS Number-67-56-1, methyl hydroxide, methylol, pyroxylic spirit, carbinol, wood spirit, wood naphtha, wood alcohol, colonial spirit, columbian spirit, methyl alcohol, 1L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
67-56-1 | |
MFCD00004595 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
64.7°C | |
-98°C | |
Amber glass bottle | |
128 hPa at 20°C |
CH4O | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
Liquid | |
Colorless | |
500mL | |
1.11 | |
0.55cp at 20°C |
Safety and Handling
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.