missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4,

Product Code. 16265880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
25 mL
50 mL
Unit Size:
10mL
25mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16265880 1 mL 10mL
16327731 10 mL 10mL
16275880 25 mL 25mL
16398196 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16265880 Supplier Thermo Scientific Acros Supplier No. 464040100
  • ONLINE EXCLUSIVE PRICE
    £101.00
    £97.90 / 10mL
    Save £3.10
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CD4O | 36.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CD4O
Molecular Weight (g/mol) 36.06

Specifications

Density 0.8880 g/mL
Flash Point 12°C
Refractive Index 1.3262
Specific Gravity 0.8880
Quantity 1 mL
Melting Point -99.0°C
Boiling Point 65.0°C
CAS 811-98-3
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Grade NMR
Chemical Name or Material Methanol-d4
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol-d4, for NMR, 99.8 atom % D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.