missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4, for NMR, 99.8 atom % D

Product Code. p-3734303 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 mL
25 mL
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10514711

Brand: Thermo Scientific Acros 321280100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 811-98-3 | CH4O | 36.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568

Specifications

Physical Form Liquid
Color Colorless
Density 0.8880g/mL
Deuterium (D) 99.75% min.
Flash Point 12°C
Packaging Glass Bottle
Refractive Index 1.3262
Water 0.05% max. (Coulometric)
Quantity 10 mL
Melting Point -99.0°C
Boiling Point 65.0°C
Linear Formula CD3OD
Beilstein 01, III, 1187
SMILES CO
IUPAC Name trideuterio(deuteriooxy)methane
Molecular Weight (g/mol) 36.066
Formula Weight 36.06
Grade NMR
Chemical Name or Material Methanol-d4, 99.8 atom%D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol-d4
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Methanol-d4, Methylalcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.