missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d3, TRC

Product Code. 30474479
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474479 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30474479 Supplier Toronto Research Chemicals Supplier No. TRCM2765885G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanol-d3
CAS 1849-29-2
Molecular Formula CHD3O
Quantity 5 g
Synonym Bieleski's Solution-d3, Carbinol-d3, Methanol Cluster-d3, Methyl Alcohol-d3, Methyl-d3 Hydroxide, Methylol-d3, Monohydroxymethane-d3, NSC 85232-d3, Wood Alcohol-d3, Methan-d3-ol, Methanol (CD3OH), Methanol-1,1,1-d3, Methanol-d3, Methyl-d3 Alcohol, Trideuteriomethanol
InChI Formula InChI=1S/CH4O/c1-2/h2H,1H3/i1D3
SMILES OC([2H])([2H])[2H]
Molecular Weight (g/mol) 35.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.