missing translation for 'onlineSavingsMsg'
Learn More

Methanol-D, TRC

Product Code. 30481608
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30481608 50 g 50g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30481608

Brand: Toronto Research Chemicals TRCM27658750G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanol D (O-D)
CAS 1455-13-6
Molecular Formula C H3 D O
Quantity 50 g
Synonym Bieleski's Solution-d, Carbinol-d, Methanol Cluster-d, Methyl Alcohol-d, Methyl Hydroxide-d, Methylol-d, Monohydroxymethane-d, NSC 85232-d, Wood Alcohol-d, MeOD
InChI Formula InChI=1S/CH4O/c1-2/h2H,1H3/i2D
SMILES [2H]OC
Molecular Weight (g/mol) 33.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.