missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d, 99.5+% (Isotopic)

Product Code. 15463007
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15463007 Supplier Thermo Scientific Alfa Aesar Supplier No. 089571.14
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1455-13-6 | CH4O | 33.048 g/mol

Methanol-d is an intermediate for making other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spécification

Density 0.813 g/mL
Flash Point 11°C (54°F)
Refractive Index 1.327
Quantity 25 g
CAS 1455-13-6
Melting Point °C to 98°C
Boiling Point 64.7°C
Assay Percent Range >99.5% (Isotopic)
Molecular Formula CH4O
MDL Number MFCD00044213
Beilstein 1730948
Synonym methanol-d, methan 2h ol, methyl alcohol-d, methanol-o-d1, ch3od, deutero methanol, ch 3 od, methanol-d1, methanol d, methan-d-ol, meod
Solubility Information Completely miscible with water.
InChI Key OKKJLVBELUTLKV-VMNATFBRSA-N
SMILES CO
IUPAC Name deuteriooxymethane
Molecular Weight (g/mol) 33.048
PubChem CID 123113
Formula Weight 33.05
Chemical Name or Material Methanol-d
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.