missing translation for 'onlineSavingsMsg'
Learn More

Methanol Anhydrous, Honeywell™ Riedel de-Haen

Product Code. 15887371
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
This item is not returnable. View return policy

Product Code. 15887371

Brand: Honeywell Riedel-de Haën 66211100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contain less that 50 ppm water. Honeywell™ Riedel de-Haen Anhydrous Solvents are packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity.

  • These solvents are specially produced to a water specification of less than 50 ppm (<0.0050%).
  • Many solvents go well beyond that and are routinely produced to lower than 5 ppm (<0.0005%).
  • All packaging steps and materials are especially designed to retain solvent dryness and ensure product integrity. Before filling, amber glass bottles are oven-dried to remove all traces of water, then filled under dry nitrogen and sealed with a special cap.
  • The water content itself is strictly controlled by coulometric Karl Fischer analysis (Hydranal™) and actual results are printed on the label.
  • This line is packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.79 g/cm3 (20°C)
Quantity 100 mL
Flash Point 11°C
Packaging Septum-sealed Bottle
UN Number 1230
Percent Purity >99.9%
Chemical Name or Material Methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.