missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99.8%, for electronic use, residue free, Thermo Scientific Chemicals

Catalog Number 10133101
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO
This item is not returnable. View return policy
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For electronic use
TRUSTED_SUSTAINABILITY
Melting Point -98.0°C
Density 0.7910g/mL
Boiling Point 64.7°C
Quantity 1 L
Flash Point 12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Assay Percent Range 99.8%
Residue after Evaporation 0.0005% max.
Specific Gravity 0.791
Linear Formula CH3OH
Viscosity 0.55 mPa.s (20°C)
Beilstein 01, IV, 1227
Water 0.1% max. (Coulometric)
Fieser 11,322; 13,337
Merck Index 15, 6029
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Percent Purity 99.8%
Grade Electronic
Chemical Name or Material Methanol, Residue Free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : methanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'