missing translation for 'onlineSavingsMsg'
Learn More

Solstice Methanol

Product Code. 15955450
Change view
Click to view available options
Quantity:
7 L
18 L
45 L
Unit Size:
18 L
45 L
7 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15955450 7 L 7 L
15935450 18 L 18 L
15995650 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15955450 Supplier Solstice Supplier No. 322137L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Identificadores de productos químicos

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl alcohol
IUPAC Name methanol
SMILES CO

Especificaciones

pH Not applicable
Melting Point -98°C
Color Colorless
CAS Max % ≤100.00%
Density 0.79 g/cm3 (20°C)
Quantity 7 L
Flash Point 11°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula CH3OH
Vapor Pressure 128 hPa (20°C)
Viscosity 0.55 mPa.s (25°C)
UN Number UN1230
Beilstein 1098229
Concentration ≥99.80%
Boiling Point 64°C to 65°C (1.013 hPa)
Percent Purity ≥99.80%
Grade ACS
Chemical Name or Material Methanol
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Methanol
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
  • H370-Causes damage to organs.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.