missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonyl chloride, 99.5%

Product Code. 16274641 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
250mL
500mL
This item is not returnable. View return policy

Product Code. 16274641

Brand: Thermo Scientific Acros 125645000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-63-0 | CH3ClO2S | 114.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-63-0
Molecular Formula CH3ClO2S
Molecular Weight (g/mol) 114.54
MDL Number MFCD00007454
InChI Key QARBMVPHQWIHKH-UHFFFAOYSA-N
Synonym methanesulfuryl chloride, methyl sulfochloride, mesyl chloride, chloromethyl sulfone, chloro methyl sulfone, methanesulfonic acid chloride, methanesulphonyl chloride, mesylchloride, methylsulfonyl chloride, methyl sulfonyl chloride
PubChem CID 31297
IUPAC Name methanesulfonyl chloride
SMILES CS(Cl)(=O)=O

Specifications

Melting Point -33.0°C
Density 1.4800 g/mL
Boiling Point 161.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Linear Formula CH3SO2Cl
Refractive Index 1.4435 to 1.4455
Quantity 500 mL
Beilstein 04,5
Fieser 01,662; 02,268; 04,326; 05,435; 06,362; 07,225; 11,322; 13,176; 17,180
Merck Index 15,6027
Specific Gravity 1.48
Solubility Information Solubility aq. soln.: insoluble (reacts with water). Other solubilities: soluble in ethanol, ether, acetone and other, common organic solvents
Viscosity 1.97 mPa.s (25°C)
Formula Weight 114.55
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Methanesulfonyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methanesulphonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.