missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonyl chloride, 99.5%

Product Code. 11916993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2.5 L
250mL
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11916993 2.5 L Glass Bottle 2.5 L
10735961 100 mL Glass Bottle 100mL
10561751 250 mL Glass Bottle 250mL
16274641 500 mL Glass Bottle 500mL
10561561 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11916993 Supplier Thermo Scientific Acros Supplier No. 125640025
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 124-63-0 | CH3ClO2S | 114.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-63-0
Molecular Formula CH3ClO2S
Molecular Weight (g/mol) 114.54
MDL Number MFCD00007454
InChI Key QARBMVPHQWIHKH-UHFFFAOYSA-N
Synonym mesyl chloride, methylsulfonyl chloride, methyl sulfochloride, methanesulphonyl chloride, chloro methyl sulfone, methanesulfuryl chloride, methanesulfonic acid chloride, methyl sulfonyl chloride, chloromethyl sulfone, mesylchloride
PubChem CID 31297
IUPAC Name methanesulfonyl chloride
SMILES CS(Cl)(=O)=O

Specifications

Melting Point -33.0°C
Density 1.4800g/mL
Boiling Point 161.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass Bottle
Linear Formula CH3SO2Cl
Refractive Index 1.4435 to 1.4455
Quantity 2.5 L
UN Number 3246
Beilstein 04,5
Fieser 01,662; 02,268; 04,326; 05,435; 06,362; 07,225; 11,322; 13,176; 17,180
Merck Index 15,6027
Specific Gravity 1.48
Solubility Information Solubility in water: insoluble (reacts with water). Other solubilities: soluble in ethanol, ether, acetone and other, common organic solvents
Viscosity 1.97 mPa.s (25°C)
Formula Weight 114.55
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Methanesulfonyl Chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.