missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid (CH3SO3H), VHG™

Product Code. 30003050
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003050 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003050 Supplier VHG Labs Supplier No. VHGJMSA500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Methanesulfonic Acid (CH3SO3H)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanesulfonic Acid
CAS 75-75-2
Methods Ion Chromatography
Analyte or Component Names Methanesulfonic Acid
Shipping Condition Room Temperature
Molecular Formula C H4 O3 S
Synonym Methanesulfonic Acid, Lutropur MSA, Lutropure MAS, MCAT 1201, Mesylic acid, Methylsulfonic acid, NSC 3718, Scaleva
InChI Formula InChI=1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)
SMILES CS(=O)(=O)O
IUPAC Name methanesulfonic acid
Solvent or Matrix CH3SO3H
Molecular Weight (g/mol) 96.11
Quantity 500 g
Formula Weight 95.9881 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.