missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid, BAKER™, J.T. Baker™

Product Code. 15558814 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
0.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15558814 500 g 0.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558814 Supplier Avantor J.T.Baker Supplier No. P68407

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Specifications

CAS 75-75-2
Flash Point 189.00°C
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula CH4O3S
Quantity 500 g
Specific Gravity 1L = 1.48kg
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILES CS(=O)(=O)O
IUPAC Name methanesulfonic acid
Molecular Weight (g/mol) 96.1
PubChem CID 6395
ChEBI CHEBI:27376
Formula Weight 96.1
Chemical Name or Material Methanesulfonic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.