missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, 70% aq. soln.

Product Code. 11423398
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423398 2500 mL 2500mL
11403398 100 mL 100mL
11413398 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423398 Supplier Thermo Scientific Alfa Aesar Supplier No. A17803.0F
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-75-2 | CH4O3S | 96.1 g/mol

Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

Solubility
Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.

Notes
Light and heat sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with amines, strong reducing agents, ethyl vinyl ether, hydrofluoric acid, strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Density 1.32
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 2500 mL
UN Number UN2586
Merck Index 14,5954
Solubility Information Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.
Formula Weight 96.1
Concentration or Composition (by Analyte or Components) 70% aq. soln.
Chemical Name or Material Methanesulfonic acid
compliance-icons compliance-icons
Product Identifier
  • Methanesulphonic acid, 70% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Methanesulphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.