missing translation for 'onlineSavingsMsg'
Learn More

Methandrostenolone-d3 (1 mg/ml in Acetonitrile), TRC

Product Code. 30400453
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400453 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400453 Supplier Toronto Research Chemicals Supplier No. TRCKIT56451X1ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methandrostenolone 17-methyl-d3
CAS 869287-60-5
Molecular Formula C20 2H3 H25 O2
Quantity 1x1 mL
Synonym Androsta-1,4-dien-3-one, 17-hydroxy-17-(methyl-d3)-, (17β)-, (17β)-17-Hydroxy-17-(methyl-d3)androsta-1,4-dien-3-one, Methandienone-d3, Methandienone-D3 (17-methyl-D3), Methandrostenolone-d3 (17-methyl-d3)
InChI Formula InChI=1S/C20H28O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,12,15-17,22H,4-5,7-8,10-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1/i3D3
SMILES [2H]C([2H])([2H])[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimethyl-17-(trideuteriomethyl)-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 303.45
Formula Weight 303.2278
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.