missing translation for 'onlineSavingsMsg'
Learn More

Methallylcyclopentane, 99%, Thermo Scientific™

Catalog Number 11429117
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
219726-61-1
C9H16
124.227
MFCD00045430
AYPUQHOHNKLMOW-UHFFFAOYSA-N
methallylcyclopentane, 2-methylprop-2-en-1-yl cyclopentane, 3-cyclopentyl-2-methyl-1-propene, acmc-1caob, 2-methyl-allyl cyclopentane, 2-methyl-2-propenyl cyclopentane #, cyclopentane, 2-methyl-2-propen-1-yl
548977
2-methylprop-2-enylcyclopentane
CC(=C)CC1CCCC1
This item is not returnable. View return policy
219726-61-1
C9H16
124.227
MFCD00045430
AYPUQHOHNKLMOW-UHFFFAOYSA-N
methallylcyclopentane, 2-methylprop-2-en-1-yl cyclopentane, 3-cyclopentyl-2-methyl-1-propene, acmc-1caob, 2-methyl-allyl cyclopentane, 2-methyl-2-propenyl cyclopentane #, cyclopentane, 2-methyl-2-propen-1-yl
548977
2-methylprop-2-enylcyclopentane
CC(=C)CC1CCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3295
Formula Weight 124.23
Percent Purity 99%
Chemical Name or Material Methallylcyclopentane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'