missing translation for 'onlineSavingsMsg'
Learn More

Methacycline Hydrochloride, TRC

Product Code. 30465505
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465505 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465505 Supplier Toronto Research Chemicals Supplier No. TRCM25878550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methacycline Hydrochloride
CAS 3963-95-9
Purity Grade Notes HPLC
Molecular Formula C22 H22 N2 O8 . Cl H
Quantity 50 mg
Synonym (4S,4aR,5S,5aR,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-2-naphthacenecarboxamide hydrochloride, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride, (4S,4aR,5S,5aR,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5α,5aα,12aα)]-, 6-Demethyl-6-deoxy-5-hydroxy-6-methylenetetracycline hydrochloride, 6-Demethyl-6-deoxy-6-methylene-5-hydroxytetracycline hydrochloride, 6-Demethyl-6-deoxy-6-methyleneoxytetracycline monohydrochloride, 6-Methylene-5-hydroxytetracycline chlorohydrate, 6-Methylene-5-hydroxytetracycline hydrochloride, 6-Methyleneoxytetracycline hydrochloride, Adriamicina, Ciclobiotic, Germiciclin, Londomycin, Metacyclin hydrochloride, Metacycline hydrochloride, Metadomus, Methacycline hydrochloride, Methacycline monohydrochloride, Metilenbiotic, Optimycin, Physiomycine, Rindex, Doxycycline Monohydrate Imp. B (EP) as Hydrochloride
InChI Formula InChI=1S/C22H22N2O8.ClH/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-6,10,14-15,17,25,27-29,32H,1H2,2-3H3,(H2,23,31);1H/t10-,14-,15+,17+,22+;/m1./s1
SMILES Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@@H]3C(=C)c4cccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N
IUPAC Name (4S,4aR,5S,5aR,12aS)-4-(dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methylidene-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 478.88
Formula Weight 478.1143
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.