missing translation for 'onlineSavingsMsg'
Learn More

Methacryloyl chloride, may contain up to ca. 15% cyclic dimer, 97%, stab., Thermo Scientific Chemicals

Product Code. 11456820
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456820 25 g 25g
11466820 100 g 100g
11476820 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456820 Supplier Thermo Scientific Alfa Aesar Supplier No. L14511.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacryloyl chloride is used in the preparation of polymeric materials. It reacts with L-histidine and forms functional monomer. It is also used to prepare monomer 2-methacrylamido-caprolactam, amide monomers by amidation.

Solubility
Miscible with alcohols, ethers and organic solvents. Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-46-7
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.533
MDL Number MFCD00000716
InChI Key VHRYZQNGTZXDNX-UHFFFAOYSA-N
Synonym methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
PubChem CID 13528
IUPAC Name 2-methylprop-2-enoyl chloride
SMILES CC(=C)C(=O)Cl

Specifications

Melting Point -60°C
Density 1.07
Boiling Point 95°C to 96°C
Flash Point 2°C (35°F)
Odor Acrid
Refractive Index 1.444
Quantity 25 g
UN Number UN3488
Beilstein 878175
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohols,ethers and organic solvents. Slightly miscible with water.
Formula Weight 104.54
Percent Purity 97%
Chemical Name or Material Methacryloyl chloride, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.