missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacrylic Acid-d6, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Methacrylic Acid-d6 |
| CAS | 92140-95-9 |
| Molecular Formula | C4 D6 O2 |
| Quantity | 0.25 g |
| Synonym | 2-Propenoic-3,3-d2 acid-d, 2-(methyl-d3)- (9 CI, ACI), 2-(Methyl-d3)-2-propenoic-3,3-d2 acid-d (ACI), Methacrylic Acid-d6, Methacrylic Acid-D6, α-Methylacrylic acid-d6 |
| InChI Formula | InChI=1 S/C4H6O2/c1-3(2)4(5)6/h1H2,2H3,(H,5,6)/i1D2,2D3/hD |
| SMILES | [2 H]OC(=O)C(=C([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | deuterio 3,3-dideuterio-2-(trideuteriomethyl)prop-2-enoate |
| Molecular Weight (g/mol) | 92.1262 |
| Formula Weight | 92.0744 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction