missing translation for 'onlineSavingsMsg'
Learn More

Methacrylamide, 98%

Product Code. 11318116
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318116 500 g 500g
11308116 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318116 Supplier Thermo Scientific Alfa Aesar Supplier No. L15013.36
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-39-0 | C4H7NO | 85.106 g/mol

Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

Solubility
Soluble in water (202g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-39-0
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00008018
InChI Key FQPSGWSUVKBHSU-UHFFFAOYSA-N
Synonym methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
PubChem CID 6595
ChEBI CHEBI:51759
IUPAC Name 2-methylprop-2-enamide
SMILES CC(=C)C(=O)N

Specifications

Melting Point 107°C to 112°C
Density 1.115
Boiling Point 215°C
Quantity 500 g
Beilstein 605397
Solubility Information Soluble in water (202g/L at 20°C).
Formula Weight 85.11
Percent Purity 98%
Chemical Name or Material Methacrylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.