missing translation for 'onlineSavingsMsg'
Learn More

Methacrolein, 96%, stab. with hydroquinone, Thermo Scientific Chemicals

Catalog Number 11334798
Click to view available options
Quantity:
5 mL
25 mL
78-85-3
C4H6O
70.09
MFCD00006974
STNJBCKSHOAVAJ-UHFFFAOYSA-N
methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
6562
2-methylprop-2-enal
CC(=C)C=O
78-85-3
C4H6O
70.09
MFCD00006974
STNJBCKSHOAVAJ-UHFFFAOYSA-N
methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
6562
2-methylprop-2-enal
CC(=C)C=O

Methacrolein is used in the manufacturing of polymers, synthetic resins and plastics. It acts as a raw material of methylmalonic acid, thermoplastic fuel and in the preparation of 2,3-dibromo-2-methyl-propionaldehyde. It is also used to study the effect limonene oxidation products and the terpene oxidation product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -81°C
Color Yellow
Density 0.847 g/mL
Boiling Point 69°C to 70°C
Flash Point −15°C (5°F)
Odor Pungent
Linear Formula H2C=C(CH3)CHO
Refractive Index 1.415
Quantity 5 mL
UN Number UN2396
Sensitivity Light sensitive
Solubility Information Miscible with water,ethanol and ether.
Formula Weight 70.09
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methacrolein, Stabilized with hydroquinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'